Innovating Science® Estuary Monitoring
Catalog #: 470347-448
Supplier: Ward's Science
CAS Number:
Specifications
- Chemical method:Colorimetry
- Description:Estuary monitoring water test kit
- Parameters:Nitrogen, Potassium, Phosphorus, pH
- No. of Tests:30
Specifications
About this item
Easy way to test estuary water for multiple factors.
- 30 tests for testing estuary water samples
Test a water sample of your choice for coliform bacteria, chloride, dissolved oxygen, biochemical oxygen demand, nitrate, pH, phosphate, and temperature.
Delivery: Includes 3 water collection bottles; 3 reaction vessels; 15 pipettes; 5 titrators; 1 pack toothpicks; 1 pack pH strips; 30 phosphate powder pillows; 5 g coliform test powder; 2×30 ml starch indicator soluiton; 2×30 ml manganese chloride reagent; 2×30 ml alkaline iodide reagent; 2×30 ml sulfuric acid reagent, 50%; 2×60 ml sodium thiosulfate titrant; 60 ml Griess reagent; 0.5 g cadmium metal powder; 3×100 ml silver nitrate titrant; 3×15 ml salinity indicator solution.
Customers Who Bought This Also Bought