First Aid Central Fire Blankets, Acme United
Supplier: ACME UNITED LIMITED CA
Certificates
About this item
Fire blankets are particularly useful for smothering fat pan fires or wrapping around a person whose clothing is on fire.
- Made of fire-resistant materials
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.