Specifications
- Description:Finger pulse oximeter
- Cat. no.:CA76492-356
Specifications
About this item
Clinically tested to provide unbeatable accuracy.
- Drop tested, durable, and moisture-proof
- Simple one button operation
- Not made with natural rubber latex
- Dual-color OLED displays SpO2, PR, PI* and Pulse bar
- Low power consumption and real time battery status indication
Use to measure the oxygen level (oxygen saturation) of the blood. It is an easy, painless measure of how well oxygen is being sent to parts of the body furthest from the heart, such as the arms and legs.