First Aid Central CPR Face Shields, Acme United
Supplier: ACME UNITED LIMITED CA
Certificates
About this item
Physical protection barriers for safe mouth to mouth resuscitation.
- One-way valve filter eliminates blow back of bodily fluids to rescuer
- Great addition to any first aid kit
An ideal component for any first aid kit.
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.