Crime Cast
Catalog #: 470020-456
Supplier: FORENSICS SOURCE
Specifications
- Description:Casting frames
- Cat. no.:470020-456
- Supplier No.:4-1105
Specifications
About this item
Fast and easy casting.
- Perfect size for casting an average size shoe print
- Self contained, mix In bag
- Includes internal water bladder
Crime-Cast is a complete plaster casting mixture in a bag. This convenient, easy-to-use pouch contains everything necessary (including water) to make a plaster cast of an average-size shoe print. A small water bladder is enclosed in each Crime-Cast pouch. There’s no need to carry around water, mixing bowls and measuring cups. Simply break the internal water 'bladder', mix in the bag and pour. When dry, Crime-Cast creates a hard, white casting.
Customers Who Bought This Also Bought