About this item
This user-friendly, portable instrument offers high resolution and accurate precision measurements for pH, mV, conductivity, and oxygen in on-site applications. It can measure a maximum of 2 of these parameters simultaneously.
- Large graphic LCD with continuous LED backlight showing both pH/mV and temperature value simultaneously
- Data logging function selectable between 1 min and 1 h; memory capacity of 5000 data sets; additional 200 memory points with manual storage
- Automatic power shut-off: Adjustable from 10 min to 24 h
- Automatic buffer recognition of DIN and NIST buffers (1.68/4.00/6.86/9.18/12.54)
- Three additional technical buffers at 25 °C (4.00/7.00/10.00) in memory
- Advanced self-test and diagnostics (electrode efficiency diagnostic, drift control); calibration symbol shows when recalibration is required
- Low energy consumption: Choice of mains supply or 4× AA batteries for over 1000 operating hours
With features that include waterproof USB output for connection to a computer and a rugged IP 67 enclosure.
The MU 6100 H S2 kit (76498-142) also includes the OXY 11-3 galvanic oxygen sensor with an integrated temperature sensor (3 m cable) and a maintenance kit.
76460-522 includes meter, case, stand, 4 LR06 batteries, 50 ml beaker, short instructions, CD ROM with manual.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76498-142
- 76498-140
- 76460-522
- Model:
- MU 6100 H S2
- MU 6100 H S1
- MU 6100 H S1
- Automatic buffer recognition:
- BDH Prolabo® TITRINORM® buffers at 25 °C: 4.00/6.98/9.94
NIST/DIN 19266 (1.68/4.00/6.86/9.18/12.54)
Technical buffers at 25 °C: 4.00/7.00/10.00 - BDH Prolabo® TITRINORM® buffers at 25 °C: 4.00/6.98/9.94
NIST/DIN 19266 (1.68/4.00/6.86/9.18/12.54)
Technical buffers at 25 °C: 4.00/7.00/10.00 - BDH Prolabo® TITRINORM® buffers at 25 °C: 4.00/6.98/9.94
NIST/DIN 19266 (1.68/4.00/6.86/9.18/12.54)
Technical buffers at 25 °C: 4.00/7.00/10.00
- Conductivity accuracy:
- ±0.5% of measured value
- ±0.5% of measured value
- ±0.5% of measured value
- Conductivity cell constant:
- Depending on probe
- Depending on probe
- Depending on probe
- Temperature range:
- Auto: −5.0...+105.0; Manual: −25...+130 °C
- Auto: −5.0...+105.0; Manual: −25...+130 °C
- Auto: −5.0...+105.0; Manual: −25...+130 °C
- Temperature resolution:
- 0.1 °C
- 0.1 °C
- 0.1 °C
- Temperature accuracy:
- ±0.2 °C
- ±0.2 °C
- ±0.2 °C
- Compliance:
- IP 67/GLP
- IP 67/GLP
- IP 67/GLP