About this item
MinION™ ― your personal, portable DNA and RNA sequencer.
- Real-time sequencing of DNA or RNA — rapid results and base modification data as standard
- Sequence DNA/RNA molecules of any length — from short to ultra-long (20 bp to ≥4 Mb)
- Simple workflow, with library prep in as little as 10 minutes
- Fits in the palm of your hand; weighs just 100 g
- Connect to a laptop via USB port
Get complete control and creativity over when, where, and how often you sequence. MinION™ provides the power of nanopore sequencing in an accessible, fully portable device. Weighing only 100 g and running off a laptop, MinION™ generates tens of gigabases of real-time data in the field or lab.
The high yields of nanopore sequencing data are suitable for a range of applications, including metagenomic species identification, antibiotic resistance profiling, microbial genome assembly, plasmid sequencing, targeted sequencing, low-coverage sequencing of large genomes, and isoform-level transcriptomics.
Choose a MinION™ starter pack to suit your specific application:
- Ligation sequencing and native barcoding kits — highest output and control over read lengths (60 min. prep time; 1,000ng gDNA). Choose the latest v14 chemistry with R10.4.1 flow cells for the highest read accuracy.
- Rapid sequencing and rapid barcoding kits — fastest, simplest library preparation (from 10 min. prep time; 100ng gDNA). Choose the latest v14 chemistry with R10.4.1 flow cells for high read accuracy. These kits are able to generate sequencing accuracies of over 99% (Q20+) and duplex data but have been optimized for speed and simplicity. If users require high duplex data, we recommend the ligation sequencing kit v14 or native barcoding kit v14.
- cDNA-PCR sequencing kit — identification and quantification of full-length transcripts with highest output (~225 min. + PCR prep time; 10ng poly-A+ RNA or 500ng total RNA).
- Direct RNA sequencing kit — sequence RNA molecules directly and preserve base modifications (135 min. prep time; 300 ng poly-A+ RNA or 1 μg total RNA).
Visit the individual library prep kit web pages for more detailed information.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Related Information
Adaptive Sampling on Nanopore Technology Basecalling and methylation detection Genome assemblies and variant calling How nanopore sequencing works How Nanopore Sequencing Works
Videos
Casestudy Bacterial Genome Sequencing Casestudy Metagenomic-Sequencing Casestudy Pathogen Sequencing Large Insights into Microorganisms - Whitepaper
Application Notes
Casestudy ClosedBacterialGenomes CS 1070(EN) V1 09Jun2020 Casestudy GMplants CS 1069(EN) V1 09Jun2020 Casestudy Leukaemia CS 1080(EN) V1 15Jun2020 Casestudy OutbreakSurveillance CS 1072(EN) V1 09Jun2020 Casestudy Pathogen&AMR CS 1068(EN) V1 09Jun2020