Order Entry
ContactUsLinkComponent
Pierce™ Zinc Reversible Stain Kit, Thermo Scientific
Pierce™ Zinc Reversible Stain Kit, Thermo Scientific
Catalog #: CAPI24582
Supplier:  Thermo Scientific
CAS Number:  
Pierce™ Zinc Reversible Stain Kit, Thermo Scientific
Catalog #: CAPI24582
Supplier:  Thermo Scientific
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Pierce™ Zinc Reversible Stain Kit
  • Size:
    1.5 L
  • Cat. no.:
    CAPI24582
  • Supplier No.:
    24582

Specifications

About this item

Thermo Scientific Pierce Zinc Reversible Stain Kit provides rapid and highly sensitive detection of protein in polyacrylamide gels immediately after electrophoresis as well as erasability for subsequent transfer or band extraction.

  • Sensitive—detect 0.25ng of protein per band in a mini gel
  • Fast—the entire protocol is complete in 15 minutes
  • Easy—ready-to-use stain and developer solutions are supplied, and no fixing or washing steps are required
  • Gentle—no chemical modification of the proteins occurs; protein bands are visible because the background is stained, not the bands themselves
  • Reversible—completely erase the stain from the gel to easily recover proteins or proceed with other methods

Stains do not require any destaining and the protein bands are easily visible right in the staining tray. In addition, there is no increased background noise if overstaining occurs.

Pierce Zinc Reversible Stain Kit is an ideal option for in-gel protein detection preceding many downstream applications, because the zinc-stain is completely reversible. The white background is easily removed and the gel is restored to its original condition with the kit-supplied eraser solution or a Tris-glycine buffer. Afterwards, the proteins are free of any dyes or other contaminants and can be restained by another method, transferred to membrane for Western blotting or eluted from an excised gel piece.

Zinc-based protein gel stain provides rapid detection of proteins after their separation by SDS-PAGE. The method is as sensitive as most silver staining techniques in that it detects protein at less than 1ng per band, yet requires less than 15 minutes to complete. The simple, two-step staining protocol results in a stained gel with an opaque milky-white background and clear protein bands. The clear bands of protein are visible when the gel is placed against a dark background.

Includes: Zinc stain, developer, and eraser solutions (500 mL each)

Applications:
• Staining preceding protein recovery for antibody generation or immunological detection
• Staining before excising gel pieces for protein digestion and analysis by mass spectrometry
• Staining before electrophoretic transfer to membrane for Western blotting
• Quick purity checks before proceeding with purification or transfer for Western blotting

Customers Who Bought This Also Bought