Caps for Serum Sample Cups, Caplugs
Supplier: Caplugs
Certificates
About this item
These serum sample cup caps are designed to fit all round, conical and standard bottom serum cups.
- Non-perforated caps are appropriate for short-term storage
- Perforated caps help minimize variation caused by evaporation during processing
- Will also fit non-threaded false bottom tubes
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.