CleanMax® Clean Manufactured Sterile Frocks, Lakeland
Supplier: Lakeland
Certificates
About this item
Protective frock for use in cleanrooms and/or controlled environments.
- Manufactured clean sterile
- Zipper closure for easy donning/doffing
- With collar
- Bound seams
- Microporous film laminate material
CleanMax® products protect the cleanroom and/or controlled environment, as well as personel with packaging designed to minimize excursions and contamination.
Certifications: ASTM D6978. CE277.
Specifications
- Catalog No:
- 76478-464
- Fabric:
- Microporous film laminate
- Closure type:
- Zip closure
- Pockets:
- Without pockets
- Gender:
- Unisex
- Sleeve or leg length:
- Long
- Garment application:
- Cleanroom, Laboratory