Corepoint Scientific™ Temperature Test Chambers, Horizon Scientific
Supplier: Horizon Scientific
Certificates
About this item
These temperature test chambers provide precise temperature control for testing in laboratory settings.
- Proportional integral derivative (PID) controller with graphic display
- Temperature setpoint range: 20 to 60 °C (glass door units) or 4 to 70 °C (solid door units)
- Multi-element, independent proportional electric heating
- Condensate evaporation
- Probe access port (³/₄")
- Keyed door lock
- Natural hydrocarbon refrigerant
Units are equipped with a probe access port, audible and visual alarms, LED interior lighting, and a Proportional integral derivative (PID) controller.
All units include an 18-month parts and labor warranty plus a 5-year compressor warranty. These units run on natural, hydrocarbon refrigerant for environmental cleanliness and economic efficiency.
Certifications: ETL Listed.
Specifications
- Catalog No:
- 76491-726
- 76491-724
- 76491-722
- 76491-720
- Environmentally Preferable:
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient
- Safer Human and Environmental Health, Energy Efficient