Order Entry
ContactUsLinkComponent
Fluoro-Jade C (FJC) Powder for Identifying Degenerating Neurons, Biosesnsis
Fluoro-Jade C (FJC) Powder for Identifying Degenerating Neurons, Biosesnsis
Catalog #: 76459-464
Supplier:  Biosensis
CAS Number:  
Fluoro-Jade C (FJC), for identifying Degenerating Neurons, Form: Dry powder, Appearance: FJC visualization is accomplished using blue light or a 488 nm Laser, Excitation Peak: 495 nm, Emission Peak: 521 nm, Application: preserved tissues, including fresh-frozen, size: 30 mg
Fluoro-Jade C (FJC) Powder for Identifying Degenerating Neurons, Biosesnsis
Catalog #: 76459-464
Supplier:  Biosensis
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Fluoro-Jade C, dry powder
  • Size:
    30 mg
  • Storage conditions:
    The powdered dye can be stored desiccated at room temperature in the dark. Storage in a desiccator is recommended as FJC is hydroscopic. The 0.01% stock solution will remain stable for 3 months when stored in a refrigerator, in the dark. The 0.0001-0.0004% working solution in 0.1% acetic acid should be used within 4 hours of preparation. Diluted FJC dye solutions are not stable and should not be stored. The other diluted solutions can be reused and stored for up to 48 hours if refrigerated and protected from light. Best results require freshly diluted solutions.

    The TR-160-FJC material is shipped ambient and stable at room temperature during transport.
    The dry powder is stable for 12 months at room temperature if stored as recommended. The 0.1% stock solution can be stored at 2 - 8 °C for up to 3 months if handled aseptically. The 0.0001-0.0004% working solution should be used within 4 hours of preparation.
  • Storage temperature:
    Ambient temperature for powder.
  • Cat. no.:
    76459-464

Specifications

About this item

Fluoro-Jade C stains all degenerating neurons regardless of specific insult or mechanism of cell death. The dye is highly resistant to fading and is compatible with virtually all histological processing and staining protocols.

  • Stains all degenerating neurons regardless of specific insult or mechanism of cell death
  • Exhibits the greatest signal to background ratio, as well as the highest resolution
  • Maximal contrast and affinity for degenerating neurons
  • Identifies degenerating nerve cell bodies, distal dendrites, axons and terminals

The causes and effects of neuronal degeneration are of major interest to a wide variety of neuroscientists. Paralleling this growing interest is an increasing number of methods applicable to the detection of neuronal degeneration. Fluoro-Jade C stains all degenerating neurons regardless of specific insult or mechanism of cell death. Fluoro-Jade C exhibits the greatest signal to background ratio, as well as the highest resolution. This translates to a stain of maximal contrast and affinity for degenerating neurons. This makes it ideal for localising not only degenerating nerve cell bodies but also distal dendrites, axons and terminals. The dye is highly resistant to fading and is compatible with virtually all histological processing and staining protocols.

Following our detailed protocol, Fluoro-Jade C labels degenerating neurons which are visualised with blue light excitation, while DAPI (not included) counter stains cell nuclei, visualised with ultra-violet illumination. The Fluoro-Jade C dye can be used on all kinds of preserved tissues, including fresh-frozen, paraformaldehyde or formalin fixed, and formalin fixed, paraffin-embedded tissues.

FJC visualization is accomplished using blue light or a 488 nm Laser. Excitation Peak: 495 nm; Emission Peak: 521 nm; Filter system for visualizing: Fluorescein/FITC.

Specificity: Degenerating neurons, and neuronal degeneration. There is no specific staining in normal healthy brain.

Cross reactivity: Some researchers under some conditions report blood vessel staining with Fluoro Jade. This may be because Fluoro Jade is an analogue of eosin (which stains blood cells). In general, good perfusion and preparation of the tissue should help prevent blood vessel staining but it may not be possible to eliminate it entirely. In our experience it is generally possible to distinguish neuronal from blood vessels staining by eye.

Delivery: Materials supplied:30 mg Fluoro-Jade C, dry powder and detailed protocol.
Caution: Details of reconstitution of the FJC and a full list of additional reagents and equipment required for use are detailed in the protocol.- see Related Documents.
Customers Who Bought This Also Bought