Calcium chloride dihydrate 99.0-103.0% (by complexometric analysis), crystalline Ph. Eur., JP, USP, FCC, E509, SAFC®
Catalog #: CA1.01867.9025
Supplier: EMD MILLIPORE – PCS CA EXCEP
CAS Number: 10035-04-8
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Packaging:Plastic sack
- Size:25 kg
- Cat. no.:CA1.01867.9025
- Supplier No.:1.01867.9025
Specifications
Specification Test Results
- Appearance:passes test
- Assay (complexometric):99.0 - 103.0 %
- Identity:passes test
- Appearance of solution (10 %; water):passes test
- Acidity or alkalinity:passes test
- pH-value (5 %; water):4.5 - 9.2
- Fluoride (F):≤ 0.004 %
- Hypochlorite (ClO):passes test
- Sulfate (SO₄):≤ 0.024 %
- Heavy metals (as Pb):≤ 0.0005 %
- Al (Aluminium):≤ 0.00002 %
- As (Arsenic):≤ 0.00015 %
- Ba (Barium):passes test
- Fe (Iron):≤ 0.0010 %
- Hg (Mercury):≤ 0.0001 %
- Pb (Lead):≤ 0.0002 %
- Magnesium and alkali metals:≤ 0.5 %
- Substances precipitated by ammonia (Al,Fe,PO₄):passes test
- Residual solvents (Ph Eur/USP/ICH):excluded by manufacturing process
- Elemental impurity specifications have been set considering ICH Q3D (Guideline for Elemental Impurities). Class 1-3 elements are not likely to be present above the ICH Q3D option 1 limit, unless specified and indicated (*).:
- Conforms to the purity criteria on food additives according to the current European Commission Regulation.:
- Corresponds to Ph.Eur., JP, USP, FCC, E509:
- SAFC®, Calcium chloride dihydrate 99.0-103.0% (by complexometric analysis), crystalline Ph. Eur., JP, USP, FCC, E509:
Specification Test Results
About this item
Synonyms: Calcium Chloride Dihydrate
:Length:
:Width:
:Height:
:Ø
:Thickness:
:99,0-103,0
:M-Clarity™ quality level = MQ500 (more info.)
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 176 °C
:Flash Pt:
:Flash Pt:
:M.W. 147.01 g/mol
:Calcium chloride dihydrate
:176
:1600
:GHS07
:MQ500
Formula:CaCl₂·2H₂O
MW:147.01 g/mol
MDL Number:MFCD00149613
CAS Number:10035-04-8
Customers Who Bought This Also Bought