SP Bel-Art H-B Durac® Thermocouple Thermometer Probes, Bel-Art Products, a part of SP
Supplier: Bel-Art Products, a Part of SP
Certificates
About this item
For use with H-B Durac® Calibrated Thermocouple Thermometers or others that accept K or J type probes.
- For temperature measurement of liquids, semi-solids, or air/gasses
- Measure wide temperature ranges
- 15.24 cm (6") stainless steel probe
- Model B60901-1300 perfect for use in dewars
Customers Who Bought This Also Bought