J.T.Baker® Plate 384
Supplier: Ritter, part of Avantor
Certificates
About this item
Available in PS and PP for automated screening and storage systems.
- Alphanumeric code to assist sample identification
- Square wells for increased volume
- V-bottom for high sample recovery
- High frame (3 mm) for stable and slip-free stacking
- Higher well rims avoid cross-contamination and allow better closure with adhesive films
J.T.Baker® Plate 384 is ideal for small sample volumes.
Specifications
- Catalog No:
- 43004-0120
- 43004-0200
- Environmentally Preferable:
- Low Manufacturing Impact
- Low Manufacturing Impact
- Environmental certification:
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)
- ISO 50001 (Certificate Registration No. 10000406726-MSC-RvA-DEU)