VWR® Vertical Freezer Racks, Stainless Steel
Supplier: VWR International
Certificates
About this item
Stainless steel vertical rack for 2" boxes.
- Double welds
- Smoothed edges
- Lifetime warranty
These freezer racks are made to the highest levels of quality with pharmaceutical grade stainless steel, double welds, smoothed edges and an unconditional lifetime warranty.
Customers Who Bought This Also Bought