Buffer, EDTA Buffer Solution pH 8,5 (0,5 mol/L)
Catalog #: CA82023-102
Supplier: G-Biosciences
CAS Number:
Specifications
- Description:EDTA, 0.5 M
- Size:100 ml
- Cat. no.:CA82023-102
- Supplier No.:R013
Specifications
About this item
A DNase and RNase free solution of 0.5M EDTA.
Customers Who Bought This Also Bought