Ammonium acetate solution 5M
Catalog #: CA82023-100
Supplier: G-Biosciences
CAS Number: 631-61-8
Specifications
- Size:100 ml
- Supplier No.:R012
Specifications
About this item
Synonyms: Acetic acid ammonium salt
:Storage temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: 114 °C
:Flash Pt:
:Flash Pt:
:M.W. 77.08 g/mol
:114
:136
Formula:H₃CCOONH₄
MW:77.08 g/mol
Storage Temperature:Ambient
MDL Number:MFCD00013066
CAS Number:631-61-8
Customers Who Bought This Also Bought