Manual/Press, Model 3626, Spex SamplePrep
Catalog #: 76182-404
Supplier: SPEX
Specifications
- Description:Press
- W×D×H:51×20×26 cm (20×8×10¼")
- Weight:63.5 kg (115 lbs.)
- Cat. no.:76182-404
- Electrical:230 V, 50 Hz
Specifications
About this item
Bench top 12-ton (10.9 metric ton) Carver® hydraulic laboratory pellet press that accepts 13 mm pellet die sets.
- Accepts SPEX SamplePrep 13 mm evacuable pellet die set
- Can be operated anywhere in the world, even under the most primitive conditions
- Economical alternative to motorized presses for labs with a small sample load
- Hardened steel buffer plate is engraved with concentric circlesand enables easy centering of the die
- Manual, hydraulic laboratory press with buffer plate
- Rugged, reliable and noted for its extraordinary durability
- Upper platen is equipped with a safety shield for operator protection
Manually controlled. Ideal for pressing sample pellets for IR.
Certifications: CE approved.
Customers Who Bought This Also Bought