To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- pH:7.25 - 7.55
- Description:VWR® TBS-T Buffer Powder
- Size:100 pks
- Cat. no.:76371-740
- Formulation:137 mM NaCl, 2.7 mM KCl, 50 mM Tris, 0.1% Tween-20
Specifications
About this item
VWR® Buffer Powder Packs can save time and money, as there is no need to measure and weight many separate chemical components which is required when making buffers from scratch.
- Buffer powder quickly dissolves to make 1L of 1X solution
- No weighing or measuring of separate components
- No pH adjustments required
- Economical shipping, long shelf life
- Quality and consistency guaranteed
VWR® Pre-measured buffer packs are extremely easy to use and are formulated so each pack makes 1 Liter of 1X buffer. Simply tear open a pack and mix completely with 1 liter of distilled water.
Shipping costs are also minimal, as each case of 50 or 100 packs is compact and weighs only 2.5 Lbs.