Specifications
- Description:Linearity FD General Chemistry Panel 3 for Abbott systems
- Size:1 ml
- Cat. no.:76480-192
- Includes:5 vials
Specifications
About this item
The Audit® MicroControls™ Linearity FD General Chemistry Panel 3 for Abbott Systems is intended to simulate human patient samples for use as assayed quality control material, determining linearity, calibration verification, and the verification of reportable range for the DBIL and TBIL analytes.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.