Specifications
- Description:Linearity FD General Chemistry Panel 1 for Abbott systems
- Size:3 ml
- Cat. no.:76480-188
- Includes:10 vials
Specifications
About this item
It is intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for BUN, calcium, chloride, CO₂, creatinine, glucose, iron, lactate, magnesium, phosphate, potassium, sodium, and uric acid.
Five distinct levels that demonstrate a linear relationship to each other for their respective analytes, reagents and instruments.