Specifications
- Format:5 vials
- Description:Linearity FLQ ISE + Trigs for Siemens Dimension
- Cat. no.:76479-182
- Volume:3 ml
Specifications
About this item
The Audit® MicroControls™ Linearity FLQ ISE + Trigs for Siemens Dimension is intended to simulate human patient samples for use in determining linearity, calibration verification, and the verification of reportable range for the following analytes: chloride, potassium, sodium and triglycerides.