Specifications
- Description:Linearity FD iPTH for Ortho vitros, 5 vials
- Cat. no.:76479-170
- Capacity:3 ml
Specifications
About this item
Intended to simulate human patient serum samples for purpose of determining linearity, calibration verification and verification of reportable range for iPTH.
The Linearity FD iPTH for Ortho Vitros is a five level linearity set. Each level contains the following analytes: iPTH. These five levels demonstrate a linear relationship to each other for their respective analytes.