Eisco LabGlass® Distilling Flasks with Oblique Neck, Round Bottom, Interchangable Joints
Supplier: Wards
Certificates
About this item
High quality round bottom distilling flasks with oblique neck.
- Made of high quality borosilicate glass 3.3
- Interchangable, ground joints
- Oblique neck design
- Round bottom
- Autoclavable
- Superior chemical durability
- Low temperature gradient - can withstand high temperatures and thermal shocks
- Heat tolerances: Strain point 515 °C; annealing point 565 °C and softening point 820 °C
Eisco manufactures all LabGlass products using state-of-the-art equipment, and following the highest standards of quality control. Each product is carefully designed by a team of laboratory professionals and experienced engineers with quality and durability in mind. They are manufactured with only the highest grade, raw materials and according to international quality standards to ensure that they can withstand use in even the toughest laboratory environments.
Customers Who Bought This Also Bought