Order Entry
ContactUsLinkComponent
His 2-Step Purification Protocol Tagged Column Package, Cytiva
His 2-Step Purification Protocol Tagged Column Package, Cytiva
Catalog #: CA76460-358
Supplier:  Cytiva
Tagged column package, His 2-step purification protocol, includes affinity chromatography and size chromatography columns to achieve high purity of his-tagged protein using a 2-step purification protocol, prepacked with Ni Sepharose High Performance (HP) affinity resin for preparative purification
His 2-Step Purification Protocol Tagged Column Package, Cytiva
Catalog #: CA76460-358
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Tagged column package, His (2-step purification protocol)
  • Storage:
    4...30 °C
  • Cat. no.:
    CA76460-358
  • Sample Volume:
    5 ml

Specifications

About this item

Column package includes affinity chromatography and size exclusion chromatography columns to achieve very high purity of his-tagged proteins using a 2-step purification protocol.

  • Contains a HisTrap HP column 1 ml prepacked with Ni Sepharose High Performance (HP) affinity resin for preparative purification of his-tagged proteins
  • Final purity achieved with HiPrep 16/60 Sephacryl S-200 HR size exclusion chromatography column, removes the trace of impurities or closely related substances and imidazole
  • Includes instructions with guidance and tips showing how to get started

Ni Sepharose HP affinity resin consists of highly cross-linked agarose beads to which a chelating group has been coupled. This chelating group is precharged with nickel, which selectively retains proteins with exposed histidine groups. Ni Sepharose HP has a binding capacity that exceeds 40 mg/ml, resulting in high protein recovery.

The size exclusion column is prepacked with Sephacryl S-200 HR chromatography resin, which has a broad fractionation range of Mr 5000 to ~ 250000 for globular proteins. The high sample capacity of the HiPrep column.

Customers Who Bought This Also Bought