Drop-Boy Dropper Bottle
Catalog #: 470332-512
Supplier: Dynalon
Specifications
- Capacity:50 ml
- Description:Drop-Boy dropper bottle
- Cat. no.:470332-512
Specifications
About this item
Precise drop dosing.
- Bottle should have dropper tip cut off before first use
- Reclosable using screw-on cap
Drop-Boy is a HDPE dropping bottle with a LDPE drop dispenser and separate polypropylene screw cap.
Customers Who Bought This Also Bought