Mini Plant Press
Supplier: Ward's Science
Certificates
About this item
For hobbyist or field collection.
- Sized for easy portability
- Elastic bands provide even compression
The solution to the field ecologist's or naturalist's problem of bringing back specimens for identification from the field. These very portable yet extremely functional mini-presses should be in every field biologists kit. As a teaching tool and for student use the mini-press is ideal.
Delivery: Plant press has two masonite boards, 12 cards and driers and two elastic bands.
Customers Who Bought This Also Bought