Order Entry
ContactUsLinkComponent
Modern Protein A Antibody Column Package (2-Step Purification Protocol), Cytiva
Modern Protein A Antibody Column Package (2-Step Purification Protocol), Cytiva
Catalog #: CA76460-356
Supplier:  Cytiva
Antibody package, modern Protein A (2-step protocol), Product type: Liquid, Color: White to yellowish, Odor: Alcohol-like[Slight], Odor threshold: 180 ppm, Flash point Closed cup: 38 to 43 degree C, Solubility: Easily soluble in the following materials: cold water and hot water
Modern Protein A Antibody Column Package (2-Step Purification Protocol), Cytiva
Catalog #: CA76460-356
Supplier:  Cytiva
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    Antibody column package, modern Protein A (2-step purification protocol)
  • Storage:
    2...8 °C
  • Cat. no.:
    CA76460-356
  • Sample Volume:
    5 ml

Specifications

About this item

The Antibody column package includes modern protein A affinity and size exclusion chromatography columns to obtain highly pure human IgG.

  • Contains columns suitable for a successful 2-step protocol of human antibodies: HiTrap MabSelect PrismA (1×1 ml) and HiPrep 16/60 Sephacryl S-300 HR
  • Includes instruction with guidance and tips to help you quickly get started

HiTrap MabSelect PrismA column is prepacked with MabSelect PrismA resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant, providing excellent purity in one step. Obtain final high purity with the HiPrep 16/60 Sephacryl S-300 HR size exclusion column, which removes antibody aggregates and/or fragments.

MabSelect PrismA resin containing a protein A-derived ligand produced in E. coli. The ligand has been specially engineered to create an affinity resin with enhanced alkali and protease stability. The specificity of binding to the Fc region of IgG is similar to conventional Protein A and provides you with excellent purification in one step.

HiPrep 16/60 Sephacryl S-300 HR size exclusion chromatography column is prepacked with Sephacryl S-300 HR chromatography resin, which has a broad fractionation range of Mr 10000 to 1500000 for globular proteins. The resin is well-suited for antibody purification and the high sample capacity of the HiPrep column.

Customers Who Bought This Also Bought