To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Antibody column package, modern Protein A (1-step purification protocol)
- Storage:2...8 °C
- Cat. no.:CA76460-354
- Sample Volume:0.1 - 1.5 ml
Specifications
About this item
The Antibody column package includes modern Protein A affinity resin and desalting columns for purification of human antibodies.
- Contains columns for a 1-step protocol using affinity chromatography, HiTrap MabSelect PrismA 1×1 ml and HiTrap Desalting 1×5 ml
- HiTrap MabSelect PrismA column is prepacked with resin for preparative purification of monoclonal and polyclonal antibodies from serum and cell culture supernatant
- HiTrap Desalting column is packed with Sephadex G-25 to give you quick pH neutralization of the eluted antibodies
- Includes instruction with guidance and tips to quickly get started
The package includes a HiTrap column prepacked with MabSelect PrismA resin containing a protein A-derived ligand produced in E. coli. Fermentation and subsequent purification are performed in the absence of animal products. The ligand has been specially engineered to create an affinity resin with enhanced alkali and protease stability. The specificity of binding to the Fc region of IgG is similar to that of conventional Protein A and provides excellent purification in one step.