VWR® Lab Chair Glides
Catalog #: 76473-608
Supplier: VWR International
Specifications
- Description:Glides, 2" height
- Cat. no.:76473-608
- For Use With:VWR® lab chairs
Specifications
About this item
Polyamide glides for VWR® lab chairs provide consistent seat height if using in place of casters.
- Chair remains stationary
Customers Who Bought This Also Bought