Specifications
- Capacity:90 ml
- Description:Plastic collection cup
- Cat. no.:76473-384
Specifications
About this item
The Rapid Response® Plastic collection cup is a non sterile cup made with polypropylene allowing for clarity and durability and is ideal for sample collection.
- 90 ml volume
- 45 mm diameter
- Screw-on cap
The cup includes graduations on the cup exterior for easy reading and made with polypropylene material for clarity and durability when reviewing samples. The compact size is ideal for sample testing with strips, cassettes, and 1 - 12 panel drug test.