Order Entry
ContactUsLinkComponent
Rapid Response™ Urinary Tract Infection Test Strip, BTNX
Rapid Response™ Urinary Tract Infection Test Strip, BTNX
Catalog #: 76473-526
Supplier:  BTNX INC.
CAS Number:  
Test Strip, Urinary Tract Infection, rapid, self-performing test for the detection of leukocytes and nitrite in urine, to aid in the screening of urinary tract infection (UTI), 3 per kit, Contents: 3 Test Strips, Colour Chart, Product Insert
Rapid Response™ Urinary Tract Infection Test Strip, BTNX
Catalog #: 76473-526
Supplier:  BTNX INC.
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Description:
    UTI test strip
  • Cat. no.:
    76473-526

Specifications

About this item

The Rapid Response® Urinary tract infection test strip is a rapid, self-performing test for the detection of leukocytes and nitrite in urine, to aid in the screening of urinary tract infection (UTI).

  • Sample: Urine
  • Format: Strip
  • Specificity: Leukocytes and nitrites
  • Accuracy: Leukocyte: 94.4%, Nitrite: 98.4%
  • Time to result: 1 to 2 minutes
  • Storage condition: 2 to 30 °C (36 to 86 °F)
  • Test principle: Colorimetric analysis

The test can be given to patients to check for an acute urinary tract infection, and to help monitor and assess therapy treatment.

Delivery: Includes test strips, color chart, product insert.
Caution: The Rapid Response® urinary tract infection test strip is for diagnostic use and OTC in Canada and U.S.
Customers Who Bought This Also Bought