To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:FIT test
- Cat. no.:76473-472
Specifications
About this item
The Rapid Response® Fecal immunochemical test (FIT) cassette is a rapid, qualitative test for the determination of human occult blood in feces, to aid in the early detection of gastrointestinal problems such as colorectal cancer.
- Sample: Fecal
- Format: Cassette
- Specificity: Human hemoglobin
- Time to result: 5 minutes
- Storage condition: 2 to 30 °C (36 to 86 °F)
- Test principle: Lateral flow immunoassay
Traditional guaiac-based methods to detect fecal occult blood lack sensitivity and specificity, and also have diet restrictions prior to testing. Unlike guaiac assays, the accuracy of this test is not affected by the diet of the patients.