Specifications
- Description:Liquid diptube urine controls
- Cat. no.:76473-394
Specifications
About this item
The Rapid Response® Liquid dip tube urine controls are available in two levels and are intended for use in validating the visual and analyzer reading of urinalysis, to ensure the consistent performance of Rapid Response® Urinalysis reagent strips and Rapid Response® U120 urine analyzers.
- Graduations on tubes for easy reading
- Screw-top lids to prevent leakage
For visual reading, test results are interpreted by direct comparison of the color blocks printed on the canister label. For Analyzer reading, refer to the Instruction Manual for details.