About this item
Obtain pH and dissolved oxygen readings from one meter.
- Optical DO sensor eliminates costly, tiresome membranes and solutions
- Durable waterproof and dustproof design with IP 67 rating
- Barometric pressure and salinity correction
- Memory for up to 500 data sets with USB or RS232 output to PC
- Built-in stand allows for easy benchtop use
- 500 hours battery life, or optional universal power adapter
The PD 450 meter uses optical measurement technology offering fast response times and long-term stability with minimal maintenance for dissolved oxygen measurements. Plus the automatic temperature compensation (ATC), barometric pressure compensation, and salinity correction further ensure DO measurement accuracy. The large, backlit LCD lets you view pH, mV, % saturation, or mg/L (ppm) simultaneously with temperature.
Rugged, IP 67-rated waterproof housing makes the meter ideal for plant or field testing. Additional features include ready (stability) indicator, smart averaging, hold function, calibration alarm, and battery-life indicator. Perform data logging with manual or timed collection of up to 500 data sets with real-time clock time/date stamp.
Automatic buffer recognition: DIN, NIST, USA, and custom
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Catalog No:
- 76221-292
- 76221-290
- DO accuracy:
- ±2%
- ±2%
- DO resolution:
- 0.1 ppm; 0.1% saturation
- 0.1 ppm; 0.1% saturation
- DO range:
- 0 - 20 ppm; 0 - 200%
- 0 - 20 ppm; 0 - 200%
- Temperature range:
- −10...110 °C (14...230 °F)
- −10...110 °C (14...230 °F)
- Dimensions:
- 3.125W×2.5" H
- 3.125W×2.5" H
- Connections:
- pH: BNC; temperature; DO: 9-Pin
- pH: BNC; temperature; DO: 9-Pin
- Calibration:
- Up to 6 points
- Up to 6 points