Order Entry
ContactUsLinkComponent
CSP Random Testing Kit, Hardy Diagnostics
CSP Random Testing Kit, Hardy Diagnostics
Catalog #: 76436-450
Supplier:  Hardy Diagnostics
CAS Number:  
CSP Random Testing Kit, contains: Five syringes of 5ml double strength Tryptic Soy Broth, 5 whirl-pak bags, 5 tests per kit, test results log sheet, Quality culture media tested for performance, pH and sterility, Kits are designed to help you follow USP <797> guidelines
CSP Random Testing Kit, Hardy Diagnostics
Catalog #: 76436-450
Supplier:  Hardy Diagnostics
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Format:
    Syringes 
  • Packaging:
    5 tests per kit
  • Description:
    CSP random testing kit
  • Size:
    25 ml
  • Cat. no.:
    76436-450
  • Fill Volume:
    5x5 ml

Specifications

About this item

A random test of CSPs (Compounded Sterile Preparations) to be performed on returned I.V. solutions. Performs five tests.

Expert technical support is available from staff of microbiologists. Quality culture media tested for performance, pH and sterility.

Certifications: Manufactured in an ISO 13485 certified facility.
Ordering information: Kit contains: Five syringes of 5 ml double strenth Tryptic Soy broth, five specimen bags, and test results log sheet.
Customers Who Bought This Also Bought