Thermo Scientific™ H-FLEX HS4 Windshield Swinging Bucket Rotor and Accessories, Thermo Scientific
Supplier: Thermo Fisher Scientific
Certificates
About this item
Specifications
- Catalog No:
- 76470-424
- 76470-422
- 76470-420
- Max. capacity:
- 16×50 ml conical tubes, 36×15 ml conical tubes, 66× 10/15 ml blood tubes, 84×5/7 ml blood tubes, 4×400 ml bottle, 10× standard microplates
- Max. RCF:
- 7197 ×g