Specifications
- Description:TriDye Ultra low range DNA ladder
- Size:1.25 ml
- Cat. no.:CA76339-754
Specifications
About this item
TriDye Ultra low range DNA ladder is a pre-mixed, ready-to-load molecular weight marker containing three tracking dyes which serve as visual aids to monitor the progress of migration during native agarose and polyacrylamide gel electrophoresis.
- Convenient, ready-to-load format
- Size range: 10 to 700 bp
- Suitable for use on both native polyacrylamide and agarose gels
- Formulated with TriDye loading dye, containing 3 different tracking dyes
The DNA ladder contains 13 bands ranging from 10 to 700 bp, most of which are generated from the digestion of a proprietary plasmid with restriction enzymes. The 50 and 300 bp bands have increased intensity to serve as reference bands. The ladder is suitable for use with for use with both native polyacrylamide and agarose gel electrophoresis.