To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Antibody type:Primary
- Antigen name:Glial fibrillary acidic protein
- Clonality:Monoclonal
- Clone:2A5
- Gene ID:Uniprot# P14136
- Host:Mouse
- Isotype:IgG1
- Antigen symbol:GFAP
- Conjugation:Unconjugated
- ImmunoChemistry:Yes
- ImmunoFluorescence:Yes
- Size:100 µg
- Western blot:Yes
- Epitope:The epitope for this antibody has been mapped to the N-terminal region of the alpha-helical coiled-coil rod-region of human GFAP isotype I, specifically within amino acids 71-217.
- Form:Lyophilized
- Antigen synonyms:Glial fibrillary acidic protein, GFAP
- Shipping temperature:RT
- Immunogen:GFAP isolated biochemically from pig spinal cord was used as the immunogen.
- Purification:Affinity-purified
- Cat. no.:76459-466
Specifications
About this item
GFAP is a 50 kDa intra-cytoplasmic filamentous protein of the cytoskeleton in astrocytes. During the development of the central nervous system, it is a cell-specific marker that distinguishes astrocytes from other glial cells. GFAP immunoreactivity has been shown in immature oligodendrocytes, epiglottic cartilage, pituicytes, papillary meningiomas, myoepithelial cells of the breast and in non-CNS: Schwann cells, salivary gland neoplasms, enteric glia cells, and metastasizing renal carcinomas.
Applications: This antibody is recommended for use in immunohistochemistry (1:500 - 1:1,000) and western blotting (1:1,000 - 1:2,000). Biosensis recommends optimal dilutions/concentrations should be determined by the end user.
Type: Primary
Antigen: GFAP
Clonality: Monoclonal
Clone: 2A5
Conjugation: Unconjugated
Epitope: The epitope for this antibody has been mapped to the N-terminal region of the alpha-helical coiled-coil rod-region of human GFAP isotype I, specifically within amino acids 71-217.
Host: Mouse
Isotype: IgG1
Reactivity: Pig, human, mouse, rat, cow