Order Entry
ContactUsLinkComponent
Endoglycoceramidase I (EGCase I)
Endoglycoceramidase I (EGCase I)
Catalog #: 76196-404
CAS Number:  
Endoglycoceramidase I (EGCase I) 150 milliunits
Endoglycoceramidase I (EGCase I)
Catalog #: 76196-404
CAS Number:  
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.

To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:

  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address

* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.

Specifications

  • Size:
    150 milliunits
  • Enzyme name:
    Endoglycoceramidase I (EGCase I)
  • Concentration:
    6 units/ml
  • Cat. no.:
    76196-404

Specifications

About this item

Endoglycoceramidase I (EGCase I) catalyzes the hydrolysis of the β-glycosidic linkage between oligosaccharides and ceramides in various glycosphingolipids.

  • Cleaves the β-glycosidic linkage between oligosaccharides and ceramides in glycosphingolipids
  • Oligosaccharide and ceramide remain intact, leaving them suitable for further analysis
  • This is an Enzyme for Innovation (EFI), a project initiated by NEB to provide unique enzymes

The enzyme leaves the oligosaccharide and ceramide intact, keeping them suitable for further analysis.

Customers Who Bought This Also Bought