Specifications
- Size:50 reactions
- Cat. no.:CAMSPP-M0649S
Specifications
About this item
The Nucleoside Digestion Mix is a mixture of enzymes that provides a convenient one-step method to generate single nucleosides from DNA or RNA.
- Convenient one-step protocol
- Digests both DNA and RNA to single nucleosides
- Low-glycerol formulation significantly reduces glycerol-induced ion suppression during MS analysis
Optimized for quantitative analysis by liquid chromatography-mass spectrometry (LC-MS), this reagent eliminates the need for sequential multi-step, time-consuming digestion protocols. The Nucleoside Digestion Mix digests ssDNA, dsDNA, DNA/RNA hybrids and RNA (except mRNA cap structures) containing epigenetically modified (m5C, hm5C, f5C, ca5C, m4C, m6A, etc.), unnatural, or damaged bases. Moreover, the low-glycerol formulation (<1%) significantly reduces glycerol-induced ion suppression during mass spectrometry analysis.