Signature Series™ Flex™ Dehydrants, Epredia
Supplier: Thermo Scientific
Certificates
About this item
Epredia™ Signature Series™ Flex 100, 95 and 80 dehydrants provides gentle dehydration while demonstrtating brillant cytoplasmic staining.
- Formulation of methyl and isopropyl alcohols
- Minimizes over-dehydration in smaller specimens
- Excellent for lipid extraction, allowing for increased paraffin infiltration
- May be used as a solvent for staining applications
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.