Clear-Rite 3™ Clearant, Epredia
Supplier: Thermo Scientific
Certificates
About this item
Render complete deparaffinization and clearing during the staining process with Epredia™ Signature Series™ Clear-Rite 3™, a unique blend of isoparaffinic aliphatic hydrocarbons.
- Very low odor; benzene-free
- Use in all tissue processors and automatic stainers
- Does not produce hard or brittle specimens, even after prolonged exposure
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.