Polysorbate 80 NF, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur., J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIAL LLC
Certificates
About this item
Synonyms: Sorbitan monooleate, ethoxylated,Polyoxyethylene-80-Sorbitan Monolaurate
Melting Pt:-21 °C
Density:1.062 g/cm³
MDL Number:MFCD00082107
CAS Number:9005-65-6
:Storage temperature: Ambient
:Length:
:Width:
:Height:
:Ø
:Thickness:
:Management of Change (MOC) category = R
Flash Pt:113 °C (closed cup)
:Boiling Pt:
:Boiling Pt:
:Melting Pt:
:Melting Pt: -21 °C
:Flash Pt:
:Flash Pt: 113 °C (closed cup)
Storage Temperature:Ambient
EINECS:500-019-9
:113 °C
:M.O.C. Category R
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specification Test Results
- ChP - Identification 2:Passes Test
- ChP - Identification 3:Passes Test
- ChP - Identification 4:Passes Test
- ChP - Acidity or Alkalinity:5.0 - 7.5
- ChP - Color:Passes Test
- ChP - Ethylene glycol:≤ 0.01 %
- ChP - Diethylene glycol:≤ 0.01 %
- ChP - Ethylene oxide:≤ 0.0001 %
- ChP - Dioxane:≤ 0.001 %
- ChP - Water:≤ 3.0 %
- ChP - Freezing Test:Passes Test
- ChP - Residue on ignition:≤ 0.2 %
- ChP - Heavy Metals:≤ 0.001 %
- ChP - Arsenic (As):≤ 0.0002 %
- ChP - Fatty acid composition: oleic acid:≥ 58.0 %
- ChP - Fatty Acid Composition: myristic acid:≤ 5.0 %
- ChP - Fatty Acid Composition: palmitic acid:≤ 16.0 %
- ChP - Fatty Acid Composition: palmitoleic acid:≤ 8.0 %
- ChP - Fatty Acid Composition: stearic acid:≤ 6.0 %
- ChP - Fatty Acid Composition: linoleic acid:≤ 18.0 %
- ChP - Fatty Acid Composition: linolenic acid:≤ 4.0 %
- Additional Tests - Endotoxin Concentration (EU/mL):≤ 10
- Microbiological - Total Aerobic Microbial Count (cfu/g):≤ 100
- Microbiological - Escherichia Coli
Microbiological - Pseudomonas aeruginosa
Microbiological - Salmonella
Microbiological - Staphylococcus aureus:Passes Test - Microbiological - Total Yeast and Mold Count (cfu/g):≤ 50
- Ethylene glycol:≤ 0.10 %
- Diethylene glycol:≤ 0.10 %
- Vegetable Based:
- This product utilizes ingredients of non-animal origin and non-peanut origin.
Suitable for use in the manufacture of parenteral dosage forms: - Elemental Impurities (USP 232, EP 5.20)
Information on elemental impurities for this product is available on the associated Product:Only Class 2 (1,4 Dioxane, Ethylene Glycol) and Class 3 (acetic acid, 2-propanol) solvents are likely to be present
Class 2 solvents are below the Option 1 limits and any Class 3 solvent is <0.5%
Typical Oleic Acid Content, 77% - undefined:Regulatory Data Sheet and elemental impurity profile report
Due to the anhydrous nature of this product, sodium oleate, a carboxylate salt/soap formed naturally in the process and which can be white to brown in color, can precipitate with time and may affect product viscosity - Meets B.P. Chemical Specifications:
- Meets ChP Chemical Specifications:
- Meets E.P. Chemical Specifications:
- Meets J.P. Chemical Specifications:
- Meets N.F. Requirements:
- GMP Manufactured Product:
- CAUTION: For Manufacturing, processing or repackaging:
- Bulk Pharmaceutical Chemical:
- NF - Acid Value:≤ 2.0
- NF - Hydroxyl Value:65 - 80
- NF - Identification A:Passes Test
- NF - Identification B:Passes Test
- NF - Residue on Ignition:≤ 0.25 %
- NF - Saponification Value:45 - 55
- NF - Specific Gravity at 25°C:1.06 - 1.09
- NF - Viscosity at 25°C, cSt:300 - 500
- NF - Water (H₂O):≤ 3.0 %
- NF - Peroxide Value:≤ 10.0
- NF - Ethylene Oxide:≤ 1 ppm
- NF - Dioxane:≤ 10 ppm
- NF - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- NF - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- NF - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- NF - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- NF - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- NF - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- NF - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- EP - Acid Value:≤ 2.0
- EP - Total Ash:≤ 0.25 %
- EP - Hydroxyl Value:65 - 80
- EP - Identification A:Passes Test
- EP - Identification D:Passes Test
- EP - Peroxide Value:≤ 10.0
- EP - Ethylene Oxide:≤ 1 ppm
- EP - Dioxan:≤ 10 ppm
- EP - Saponification Value:45 - 55
- EP - Water (H₂O):≤ 3.0 %
- EP/BP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- EP/BP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- EP/BP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- EP/BP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- EP/BP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- EP/BP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- EP/BP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- JP - Acid Value:≤ 2.0
- JP - Composition of Fatty Acids - Myristic Acid:≤ 5.0 %
- JP - Composition of Fatty Acids - Palmitic Acid:≤ 16.0 %
- JP - Composition of Fatty Acids - Palmitoleic Acid:≤ 8.0 %
- JP - Composition of Fatty Acids - Stearic Acid:≤ 6.0 %
- JP - Composition of Fatty Acids - Oleic Acid:≥ 58.0 %
- JP - Composition of Fatty Acids - Linoleic Acid:≤ 18.0 %
- JP - Composition of Fatty Acids - Linolenic Acid:≤ 4.0 %
- JP - Dioxane:≤ 10 ppm
- JP - Ethylene Oxide:≤ 1 ppm
- JP - Heavy Metals (as Pb):≤ 20 ppm
- JP - Hydroxyl Value, meq KOH/g:65 - 80
- JP - Identification:Passes Test
- JP - Peroxide Value:≤ 10.0
- JP - Residue on Ignition:≤ 0.25 %
- JP - Water (H₂O):≤ 3.0 %
- JP - Saponification Value:45 - 55
- ChP - Relative Density @ 20 °C:1.06 - 1.09
- ChP - Viscosity @ 25 °C:350 - 550 mm2/s
- ChP - Acid Value:≤ 2.0
- ChP - Saponification Value:45 - 55
- ChP - Hydroxyl Value:65 - 80
- ChP - Iodine Value:18 - 24
- ChP - Peroxide Value:≤ 10
- ChP - Identification 1:Passes Test
- J.T.Baker®, Polysorbate 80 NF, Multi-Compendial, meets analytical specification of ChP, BP, JP, Ph. Eur.:
Specification Test Results
Related Information
Certificates
Search by Lot Numberspec_JT4125-03 spec_JT4125-04 spec_JT4125-06 spec_JT4125-12
Product specification
Customers Who Bought This Also Bought