About this item
The Thermo Scientific™ Megafuge ST1/ST1R Plus benchtop centrifuges offer intuitive controls and versatility for power at every turn.
- Auto-Lock rotor system
- ClickSeal™ bucket sealing system
- SMARTSpin™ technology
- Space-saving design
- Energy efficient
Secure locking system allows easy push-button installation and exchange of rotors. System saves time when switching between applications. Easy access to rotor chamber offers quick cleaning, a healthier working environment and longer unit life. Biocontainment solution seals with a snap; eliminates screw caps and complicated clips. System has glove-friendly, one-handed open/close capability. System is certified by CAMR™ in Porton Down, UK.
Advanced rotor management system maximizes acceleration, braking and residual load imbalance for each rotor and bucket option. Technology optimizes safety and improves separations.
Compact benchtop dimensions optimize working area, the ergonomic height and curved design with motorized lid latch ensure easy sample loading and unloading, easy cleaning and promotes user safety.
System offers up to 40% energy consumption savings on industry standard protocols, such as standard blood separation or conical tubes processing.
Cell Culture package includes TX400 rotor, set of four 400 ml buckets, set of four ClickSeal™ biocontainment lids, set of four 4×50 ml conical tube adapters, set of four 9×15 ml conical tube adapters.
Blood Tube package includes TX400 rotor, set of four 400ml buckets, set of four ClickSeal™ biocontainment lids, set of four 19×5/7 ml blood tube and 14×10 ml blood tube adapters.
Specifications
- Catalog No:
- CA76501-604
- CA76501-612
- CA76501-610
- 76470-442
- 76470-418
- CA76501-620
- 76470-438
- CA76501-618
- CA76501-602
- 76470-414
- CA76501-574
- 76470-454
- 76470-430
- CA76501-576
- 76470-428
- 76470-426
- Model:
- Megafuge ST1 R Plus
- Megafuge ST1 R Plus
- Megafuge ST1 Plus
- Megafuge ST1 Plus
- Megafuge ST1 R Plus
- Megafuge ST1 R Plus
- Megafuge ST1 R Plus
- Megafuge ST1 Plus
- Megafuge ST1 Plus
- Megafuge ST1 Plus
- Megafuge ST1 Plus
- Megafuge ST1 Plus
- Megafuge ST1 R Plus
- Megafuge ST1 R Plus
- Megafuge ST1 Plus
- Megafuge ST1 R Plus
- Temperature:
- –10...+40 °C
- –10...+40 °C
- –10...+40 °C
- –10...+40 °C
- –10...+40 °C
- –10...+40 °C
- –10...+40 °C
- –10...+40 °C
- Acceleration/braking profiles:
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Choice of 9 acceleration / 10 deceleration rates
- Control type:
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Microprocessor
- Display:
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Push button and high contrast LCD interface
- Voltage:
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- 120 - 240 V
- W×D×H:
- 625×660×361 mm
- 625×660×361 mm
- 445×660×362 mm
- 445×660×362 mm
- 625×660×361 mm
- 625×660×361 mm
- 625×660×361 mm
- 445×660×362 mm
- 445×660×362 mm
- 445×660×362 mm
- 445×660×362 mm
- 445×660×362 mm
- 625×660×361 mm
- 625×660×361 mm
- 445×660×362 mm
- 625×660×361 mm