To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Temperature Range:Ambient 65 °C (reagent temperature)
- Description:Rapid tissue processor
- Cat. no.:76421-772
- W×D×H:30×25×26"
Specifications
About this item
HistoPro® 600 provides continuous load, rapid tissue processing without microwave artifacts.
- No microwave artifacts
- Load up to 100 cassettes every 15 minutes
- Over 2500 cassettes in an 8-hour shift
- Compact footprint
- Built-in fume extraction can connect to lab fume exhaust system
- Runs on regular power outlet
- Stir bar and basket agitation
HistoPro® 600 is compact, cost conscious package and easy to use. Processing mode features rotating baskets, continuous load, and programmable station time. Fume control: Activated charcoal filter and connection to external fume hood.
Capacity: 6 Stations; 1 load station; Up to 5 baskets simultaneously (1.8 L reagent capacity)
Display: 20 characters × 8 lines LCD
Programmable: Station times 1 min. to 12 hours
Vacuum: 5 psi (25 cm of Hg) below ambient