I2456 Cut Resistant Impact Gloves, Wells Lamont
Supplier: Wells Lamont
Certificates
About this item
Lightweight glove delivers great cut protection.
- Hi-Vis
- TPR impact protection
- Reinforced thumb crotch
It provides protection for a wide range of applications with impact and cut hazards. The lightweight liner also provides great dexterity and comfort along with the padded palm for added protection. The TPR on the back-of-hand and the fingers is great for deflecting and absorbing impact as well. A sandy nitrile coating on the palm provides superior grip in oily applications.
Certifications: ANSI cut A5, abrasion level 5, impact level 2, puncture level 5.
Ordering information: Sold by the dozen pairs.
Specifications
- Catalog No:
- 76451-966
- 76451-968
- 76451-974
- 76451-970
- 76451-972
- Ambidextrous/Hand-specific:
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Hand-specific
- Disposable / reusable:
- Reusable
- Reusable
- Reusable
- Reusable
- Reusable
- Latex/Latex-free:
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Latex-free
- Powdered/Powder-free:
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Powder-free
- Gauge:
- 13
- 13
- 13
- 13
- 13
- Cuff style:
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist
- Knit wrist