Specifications
- Description:Monarch® HMW gDNA tissue lysis buffer
- Cat. no.:CA76470-772
- Volume:62 ml
Specifications
About this item
The Monarch® HMW gDNA tissue lysis buffer, supplied as a component of the Monarch HMW DNA extraction kit for tissue, is a highly effective lysis buffer for tissue and bacteria samples. Its composition ensures immediate and complete inactivation of nucleases in the samples at the start of the lysis.
- Supplied as a component of the Monarch HMW DNA extraction kit for tissue
- This lysis buffer enables rapid and effective lysis of various tissue and bacteria samples
- Offers maximal protection against nucleases allowing for the isolation of intact HMW DNA