Monarch® gDNA Elution Buffer II, New England Biolabs
Catalog #: CA76470-770
Supplier: New England Biolabs (NEB)
CAS Number:
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
- • State issued document with your organization's Federal Tax ID Number
- • State issued document with your organization's Resale Tax ID Number
- • City or County issued Business License
- • State Department of Health Services License
- • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Specifications
- Description:Monarch® gDNA elution buffer II
- Size:24 ml
- Cat. no.:CA76470-770
Specifications
About this item
The Monarch® gDNA elution buffer II is supplied as a component of the Monarch HMW DNA extraction kits.
- Supplied as a component of the Monarch HMW DNA extraction kits
- Optimized for long term storage of HMW gDNA
- 10 mM Tris, pH 9.0, 0.5 mM EDTA
Delivery: Supplied as 2 bottles of 12 ml.
Customers Who Bought This Also Bought